Assay Details
Target Gene Details
Entrez Gene ID: | 320118 |
Gene Name: | F-box and leucine-rich repeat protein 13 |
Gene Aliases: |
4921539K22Rik |
Location: |
Chr.5:21483847-21645698 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fbxl13 | NM_001199632.1 | NP_001186561.1 | ||
| NM_177076.4 | NP_796050.2 | |||
| AK029566.1 | BAC26515.1 | |||
| AK030172.1 | BAC26821.1 | |||
| BC145291.1 | ||||
| BC145292.1 |

Genomic Map