Assay Details
Target Gene Details
Entrez Gene ID: | 64339 |
Gene Name: | fibronectin type III domain containing 4 |
Gene Aliases: |
2810430J06Rik, 6330410H20Rik, AB030187, AI838506, AW487863, FRCP1, Fnmp1 |
Location: |
Chr.5:31292245-31295877 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fndc4 | NM_022424.5 | NP_071869.2 | ||
| AB030187.1 | BAA92751.1 | |||
| AK013203.1 | BAB28710.2 | |||
| AK018154.1 | ||||
| AK163183.1 | BAE37223.1 | |||
| BC027164.1 | AAH27164.1 | |||
| BC139278.1 | ||||
| BC139292.1 |

Genomic Map