Assay Details
Target Gene Details
Entrez Gene ID: | 71176 |
Gene Name: | F-box protein 24 |
Gene Aliases: |
4933422D21Rik, Fbx24 |
Location: |
Chr.5:137612505-137625584 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fbxo24 | NM_027708.1 | 3 | 328 | NP_081984.1 |
| AK016870.1 | 3 | 329 | BAB30476.1 |

Genomic Map