Assay Details
Target Gene Details
Entrez Gene ID: | 23950 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily B, member 6 |
Gene Aliases: |
Mrj, mDj4 |
Location: |
Chr.5:29735687-29786478 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajb6 | NM_001037940.4 | NP_001033029.1 | ||
| NM_001127367.1 | NP_001120839.1 | |||
| NM_011847.4 | NP_035977.2 | |||
| AB028854.1 | BAA88302.1 | |||
| AF035962.1 | AAC16759.1 | |||
| AK032039.1 | ||||
| AK133805.1 | BAE21853.1 | |||
| AK134387.1 | BAE22124.1 | |||
| AK144182.1 | BAE25752.1 | |||
| AK151219.1 | BAE30213.1 | |||
| AK151752.1 | BAE30661.1 | |||
| AK152626.1 | BAE31369.1 | |||
| AK152632.1 | BAE31375.1 | |||
| AK153091.1 | BAE31714.1 | |||
| AK169332.1 | BAE41085.1 | |||
| AK169767.1 | BAE41354.1 | |||
| BC003702.1 | AAH03702.1 | |||
| BC083349.1 | AAH83349.1 |

Genomic Map