Assay Details
Target Gene Details
Entrez Gene ID: | 108069 |
Gene Name: | glutamate receptor, metabotropic 3 |
Gene Aliases: |
0710001G23Rik, Gprc1c, mGlu3, mGluR3 |
Location: |
Chr.5:9485236-9725352 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grm3 | NM_181850.2 | 5 | 2844 | NP_862898.1 |
| AK002958.1 | 2 | 1048 | ||
| BC060060.1 | 3 | 1202 | AAH60060.1 | |
| BC111891.1 | 5 | 2662 | ||
| BC113170.1 | 5 | 2662 |
More Information
Set Membership: |
|

Genomic Map