Assay Details
Target Gene Details
Entrez Gene ID: | 140709 |
Gene Name: | collagen, type XXVI, alpha 1 |
Gene Aliases: |
9430032K24Rik, BC002218, Col26a, Emid2, Emu2 |
Location: |
Chr.5:136741759-136883376 on Build GRCm38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Col26a1 | NM_024474.2 | NP_077794.2 | ||
| AB085837.1 | BAB96760.1 | |||
| AJ416092.1 | CAC94779.1 | |||
| AK076445.1 | ||||
| AK133882.1 | ||||
| AK143515.1 | ||||
| BC002218.1 | AAH02218.1 | |||
| BC075713.1 | AAH75713.1 |
More Information
Set Membership: |
|

Genomic Map