Assay Details
Target Gene Details
Entrez Gene ID: | 140709 |
Gene Name: | collagen, type XXVI, alpha 1 |
Gene Aliases: |
9430032K24Rik, BC002218, Col26a, Emid2, Emu2 |
Location: |
Chr.5:136741759-136883376 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Col26a1 | NM_024474.2 | 12 | 1227 | NP_077794.2 |
| AB085837.1 | 12 | 1298 | BAB96760.1 | |
| AJ416092.1 | 12 | 1227 | CAC94779.1 | |
| AK076445.1 | 12 | 1242 | ||
| AK133882.1 | 12 | 1246 | ||
| AK143515.1 | 12 | 1280 | ||
| BC002218.1 | 5 | 263 | AAH02218.1 | |
| BC075713.1 | 12 | 1241 | AAH75713.1 |
More Information
Set Membership: |
|

Genomic Map