Assay Details
Target Gene Details
Entrez Gene ID: | 13801 |
Gene Name: | enamelin |
Gene Aliases: |
abte |
Location: |
Chr.5:88487975-88506049 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Enam | NM_017468.3 | NP_059496.1 | ||
| BC115848.1 | ||||
| U82698.1 | AAB94312.1 |
More Information
Set Membership: |
|

Genomic Map