Assay Details
Target Gene Details
Entrez Gene ID: | 242851 |
Gene Name: | guanine nucleotide binding protein, alpha transducing 3 |
Gene Aliases: |
Ggust, Gtn |
Location: |
Chr.5:17962570-18019668 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gnat3 | NM_001081143.1 | NP_001074612.1 | ||
| AK040065.1 | BAE20568.1 | |||
| BC147449.1 | ||||
| BC147466.1 | ||||
| BC147839.1 | ||||
| BC147841.1 |
More Information
Set Membership: |
|

Genomic Map