Assay Details
Target Gene Details
Entrez Gene ID: | 14693 |
Gene Name: | guanine nucleotide binding protein (G protein), beta 2 |
Gene Aliases: |
Gnb-2 |
Location: |
Chr.5:137528127-137533229 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gnb2 | NM_010312.4 | NP_034442.1 | ||
| AK002604.1 | ||||
| AK146941.1 | BAE27550.1 | |||
| AK151628.1 | BAE30562.1 | |||
| BC029077.1 | AAH29077.1 | |||
| BC059942.1 | AAH59942.1 | |||
| BC062178.1 | AAH62178.1 | |||
| U34960.1 | AAC72250.1 | |||
| U38505.1 | AAB01736.1 |
More Information
Set Membership: |
|

Genomic Map