Assay Details
Target Gene Details
Entrez Gene ID: | 215114 |
Gene Name: | huntingtin interacting protein 1 |
Gene Aliases: |
2610109B09Rik, A930014B11Rik, E130315I21Rik, mKIAA4113 |
Location: |
Chr.5:135406518-135545319 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hip1 | NM_146001.2 | NP_666113.2 | ||
| AK020853.1 | 1 | 927 | BAB32229.1 | |
| AK147246.1 | ||||
| AK147269.1 | ||||
| AK220182.1 | BAD90367.1 | |||
| BC017516.1 | AAH17516.1 |
More Information
Set Membership: |
|

Genomic Map