Assay Details
Target Gene Details
Entrez Gene ID: | 13406 |
Gene Name: | dentin matrix protein 1 |
Gene Aliases: |
AV020965, Dmp, PP |
Location: |
Chr.5:104197558-104214109 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dmp1 | NM_016779.2 | NP_058059.2 | ||
| AK132177.1 | BAE21014.1 | |||
| AK157973.1 | BAE34293.1 | |||
| BC113753.1 | AAI13754.1 | |||
| U65020.1 | AAB93764.1 |

Genomic Map