Assay Details
Target Gene Details
Entrez Gene ID: | 70681 |
Gene Name: | family with sequence similarity 175, member A |
Gene Aliases: |
3830405G04Rik, 5630400M01Rik, AI506069, AL024423, AV118690, Ccdc98 |
Location: |
Chr.5:100804802-100845216 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam175a | NM_172405.3 | NP_765993.1 | ||
| AK014420.1 | BAC25434.1 | |||
| AK042001.1 | BAC31129.1 | |||
| AK042339.1 | BAC31229.1 | |||
| AK051816.1 | BAC34780.1 | |||
| AK167540.1 | BAE39607.1 | |||
| BC029845.1 | AAH29845.1 |

Genomic Map