Assay Details
Target Gene Details
Entrez Gene ID: | 207704 |
Gene Name: | GTP-binding protein 10 (putative) |
Gene Aliases: |
4930545J22Rik, BC034507 |
Location: |
Chr.5:5537450-5559774 on Build GRCm38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtpbp10 | NM_153116.1 | NP_694756.1 | ||
| AK139595.1 | BAE24078.1 | |||
| AK160252.1 | ||||
| BC034507.1 | AAH34507.1 |
More Information
Set Membership: |
|

Genomic Map