Assay Details
Target Gene Details
Entrez Gene ID: | 11991 |
Gene Name: | heterogeneous nuclear ribonucleoprotein D |
Gene Aliases: |
Auf1, Hnrpd |
Location: |
Chr.5:99955935-99979015 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hnrnpd | NM_001077265.2 | NP_001070733.1 | ||
| NM_001077266.2 | NP_001070734.1 | |||
| NM_001077267.2 | NP_001070735.1 | |||
| NM_007516.3 | NP_031542.2 | |||
| AK077409.1 | ||||
| BC011172.1 | ||||
| BC049098.1 | AAH49098.1 | |||
| CA451481.1 | ||||
| CF751545.1 | ||||
| CX567003.1 |

Genomic Map