Assay Details
Target Gene Details
Entrez Gene ID: | 23857 |
Gene Name: | cyclin D binding myb-like transcription factor 1 |
Gene Aliases: |
Dimp, Dmp1 |
Location: |
Chr.5:9118868-9161776 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dmtf1 | NM_001110327.1 | NP_001103797.1 | ||
| NM_011806.3 | NP_035936.3 | |||
| AK029147.1 | BAC26325.1 | |||
| AK030508.1 | ||||
| AK031887.1 | BAC27593.1 | |||
| AK033208.1 | ||||
| AK039563.1 | BAC30386.1 | |||
| AK040107.1 | ||||
| AK046201.1 | BAC32635.1 | |||
| AK078015.1 | ||||
| AK145803.1 | ||||
| AK160595.1 | BAE35902.1 | |||
| AK191004.1 | ||||
| BY247901.1 | ||||
| U70017.1 | AAC52878.1 |
More Information
Set Membership: |
|

Genomic Map