Assay Details
Target Gene Details
Entrez Gene ID: | 80909 |
Gene Name: | GATS protein-like 2 |
Gene Aliases: |
7530428J21Rik, AW742413, Gats, Gatsl1 |
Location: |
Chr.5:134099748-134141782 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gatsl2 | NM_030719.3 | NP_109644.2 | ||
| AJ296173.1 | CAC14178.1 | |||
| AK039122.1 | BAC30244.1 | |||
| AK052217.1 | BAC34889.1 | |||
| BC094613.1 | AAH94613.1 |

Genomic Map