Assay Details
Target Gene Details
Entrez Gene ID: | 11991 |
Gene Name: | heterogeneous nuclear ribonucleoprotein D |
Gene Aliases: |
Auf1, Hnrpd |
Location: |
Chr.5:99955935-99979015 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hnrnpd | NM_001077265.2 | 8 | 1363 | NP_001070733.1 |
| NM_001077266.2 | 7 | 1306 | NP_001070734.1 | |
| NM_001077267.2 | 6 | 1159 | NP_001070735.1 | |
| NM_007516.3 | 7 | 1216 | NP_031542.2 | |
| AK012230.1 | 1 | 50 | ||
| AK053251.1 | 2 | 147 | ||
| AK077409.1 | 7 | 1204 | ||
| BC011172.1 | 8 | 1038 | ||
| BC049098.1 | 7 | 1216 | AAH49098.1 | |
| CA451481.1 | 6 | 644 | ||
| CF751545.1 | 4 | 367 | ||
| CX567003.1 | 4 | 338 |

Genomic Map