Assay Details
Target Gene Details
Entrez Gene ID: | 50926 |
Gene Name: | heterogeneous nuclear ribonucleoprotein D-like |
Gene Aliases: |
AA407431, AA959857, D5Ertd650e, D5Wsu145e, Hnrnpdl1, Hnrpdl, JKTBP, hnRNP DL, hnRNP-DL |
Location: |
Chr.5:100033577-100039950 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hnrnpdl | NM_016690.4 | NP_057899.2 | ||
| AB017020.1 | BAA75479.1 | |||
| AK011767.1 | ||||
| AK172629.1 | BAE43104.1 | |||
| BC021374.1 | 7 | 1001 | AAH21374.1 | |
| CX219512.1 |

Genomic Map