Assay Details
Target Gene Details
Entrez Gene ID: | 108099 |
Gene Name: | protein kinase, AMP-activated, gamma 2 non-catalytic subunit |
Gene Aliases: |
2410051C13Rik, AAKG, AAKG2, AI854673, H91620p, WPWS |
Location: |
Chr.5:24862735-25101906 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkag2 | NM_001170555.1 | 10 | 944 | NP_001164026.1 |
| NM_001170556.1 | 10 | 941 | NP_001164027.1 | |
| NM_145401.2 | 14 | 2023 | NP_663376.2 | |
| AK032238.1 | 12 | 1535 | BAC27775.1 | |
| AK034765.1 | 8 | 609 | BAC28827.1 | |
| AK154296.1 | 14 | 2024 | BAE32497.1 | |
| AK165834.1 | 10 | 883 | BAE38403.1 | |
| AY348864.1 | 10 | 874 | AAQ55224.1 | |
| BC015283.1 | 14 | 1811 | AAH15283.1 | |
| BQ572978.1 |
More Information
Set Membership: |
|

Genomic Map