Assay Details
Target Gene Details
Entrez Gene ID: | 68929 |
Gene Name: | motile sperm domain containing 3 |
Gene Aliases: |
1190005J19Rik, 5133401H10Rik, Gtig2, R124 |
Location: |
Chr.5:137596645-137601083 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mospd3 | NM_001254762.1 | NP_001241691.1 | ||
| NM_030037.2 | NP_084313.1 | |||
| AK088731.1 | BAC40534.1 | |||
| BC003880.1 | ||||
| BC039620.1 | AAH39620.1 | |||
| BE861793.1 | ||||
| BQ445267.1 | ||||
| CK617131.1 |

Genomic Map