Assay Details
Target Gene Details
Entrez Gene ID: | 68929 |
Gene Name: | motile sperm domain containing 3 |
Gene Aliases: |
1190005J19Rik, 5133401H10Rik, Gtig2, R124 |
Location: |
Chr.5:137596645-137601083 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mospd3 | NM_001254762.1 | 4 | 790 | NP_001241691.1 |
| NM_030037.2 | 5 | 943 | NP_084313.1 | |
| AK004495.1 | 2 | 188 | BAE43182.1 | |
| AK088731.1 | 4 | 781 | BAC40534.1 | |
| BC003880.1 | 4 | 800 | ||
| BC039620.1 | 5 | 924 | AAH39620.1 | |
| BE861793.1 | ||||
| BQ445267.1 | 3 | 356 | ||
| DV656015.1 | 2 | 200 |

Genomic Map