Assay Details
Target Gene Details
Entrez Gene ID: | 56874 |
Gene Name: | ring finger protein 32 |
Gene Aliases: |
1700009J01Rik, 2700025B22Rik, 4930542N22Rik, Lmbr2 |
Location: |
Chr.5:29195170-29225526 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf32 | NM_001289757.1 | 7 | 781 | NP_001276686.1 |
| NM_021470.6 | 7 | 753 | NP_067445.3 | |
| NR_110371.1 | ||||
| AF190664.1 | 4 | 515 | AAF91091.1 | |
| AK005795.1 | 7 | 708 | BAB24242.1 | |
| AK006379.1 | 7 | 697 | BAB24557.1 | |
| AK012288.1 | ||||
| AK016026.1 | 7 | 724 | ||
| BC089493.1 | 7 | 696 | AAH89493.1 |

Genomic Map