Assay Details
Target Gene Details
Entrez Gene ID: | 18127 |
Gene Name: | nitric oxide synthase 3, endothelial cell |
Gene Aliases: |
2310065A03Rik, Nos-3, eNOS, ecNOS |
Location: |
Chr.5:24364819-24384474 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nos3 | NM_008713.4 | 21 | 2763 | NP_032739.3 |
| AK077896.1 | 21 | 2765 | BAC37052.1 | |
| AK143221.1 | 20 | 2535 | ||
| AK158106.1 | 21 | 2784 | ||
| BC052636.1 | 21 | 2750 | AAH52636.1 | |
| U53142.1 | 21 | 2751 | AAC52766.1 |
More Information
Set Membership: |
|

Genomic Map