Assay Details
Target Gene Details
Entrez Gene ID: | 19353 |
Gene Name: | RAS-related C3 botulinum substrate 1 |
Gene Aliases: |
AL023026, D5Ertd559e |
Location: |
Chr.5:143505481-143528116 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rac1 | NM_009007.2 | NP_033033.1 | ||
| AK008435.1 | BAB25667.1 | |||
| AK009017.1 | BAB26027.1 | |||
| AK011072.2 | BAB69451.1 | |||
| AK034601.1 | BAC28767.1 | |||
| AK047969.1 | BAC33203.1 | |||
| AK076023.1 | BAC36128.1 | |||
| AK088825.1 | BAC40596.1 | |||
| AK166383.1 | BAE38744.1 | |||
| AK184868.1 | ||||
| AK219413.1 | ||||
| BC003828.1 | AAH03828.1 | |||
| BC051053.1 | AAH51053.1 | |||
| CX242948.1 | ||||
| JN971018.1 | ||||
| X57277.1 | CAA40545.1 |
More Information
Set Membership: |
|

Genomic Map