Assay Details
Target Gene Details
Entrez Gene ID: | 231858 |
Gene Name: | Ras association and DIL domains |
Gene Aliases: |
AI536456, D930005D10Rik |
Location: |
Chr.5:142484839-142551098 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Radil | NM_001289588.1 | NP_001276517.1 | ||
| NM_178702.4 | NP_848817.2 | |||
| AK052964.1 | BAC35223.1 | |||
| AK079539.1 | BAC37675.1 | |||
| AK083410.1 | BAC38904.1 | |||
| AK154169.1 | ||||
| AK162917.1 | ||||
| AK173277.1 | BAD32555.1 | |||
| BC056483.1 | AAH56483.1 |

Genomic Map