Assay Details
Target Gene Details
Entrez Gene ID: | 231858 |
Gene Name: | Ras association and DIL domains |
Gene Aliases: |
AI536456, D930005D10Rik |
Location: |
Chr.5:142484839-142551098 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Radil | NM_001289588.1 | 14 | 3294 | NP_001276517.1 |
| NM_178702.4 | 14 | 3225 | NP_848817.2 | |
| AI536456.1 | 2 | 503 | ||
| AK052964.1 | 14 | 3136 | BAC35223.1 | |
| AK079539.1 | 14 | 2660 | BAC37675.1 | |
| AK083410.1 | 14 | 3244 | BAC38904.1 | |
| AK154169.1 | 14 | 3171 | ||
| AK162917.1 | 14 | 3275 | ||
| AK173277.1 | 13 | 3491 | BAD32555.1 | |
| BC056483.1 | 14 | 3177 | AAH56483.1 | |
| BP764023.1 | 2 | 515 |

Genomic Map