Assay Details
Target Gene Details
Entrez Gene ID: | 18682 |
Gene Name: | phosphorylase kinase gamma 1 |
Gene Aliases: |
Phkg |
Location: |
Chr.5:129863435-129879083 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phkg1 | NM_011079.2 | 3 | 328 | NP_035209.1 |
| BC055102.1 | 3 | 328 | AAH55102.1 | |
| J03293.1 | 3 | 198 | AAA39925.1 | |
| M16216.1 | 2 | 166 | AAA39926.1 |
More Information
Set Membership: |
|

Genomic Map