Assay Details
Target Gene Details
Entrez Gene ID: | 66845 |
Gene Name: | mitochondrial ribosomal protein L33 |
Gene Aliases: |
0610009M10Rik, L33mt, MRP-L33 |
Location: |
Chr.5:31613951-31622644 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl33 | NM_025796.3 | NP_080072.2 | ||
| AB049651.1 | BAB40856.1 | |||
| AK002421.1 | BAB22088.1 | |||
| AK008431.1 | BAB25665.1 | |||
| AK021173.1 | BAB32314.1 | |||
| AK041771.1 | 1 | 633 | BAC31060.1 | |
| AK167883.1 | BAE39896.1 | |||
| BC037363.1 | AAH37363.1 | |||
| BC051471.1 | AAH51471.1 | |||
| BC055724.1 | AAH55724.1 | |||
| BC059722.1 | AAH59722.1 | |||
| BY112771.1 |
More Information
Set Membership: |
|

Genomic Map