Assay Details
Target Gene Details
Entrez Gene ID: | 68735 |
Gene Name: | mitochondrial ribosomal protein S18C |
Gene Aliases: |
1110037D14Rik |
Location: |
Chr.5:100798759-100804467 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps18c | NM_026826.1 | NP_081102.1 | ||
| AK004139.1 | BAB23188.1 | |||
| AK088290.1 | BAC40261.1 | |||
| AK145935.1 | BAE26766.1 | |||
| BC028442.1 | AAH28442.1 |
More Information
Set Membership: |
|

Genomic Map