Assay Details
Target Gene Details
Entrez Gene ID: | 18542 |
Gene Name: | procollagen C-endopeptidase enhancer protein |
Gene Aliases: |
Astt-2, Astt2, P14, PCPE-1 |
Location: |
Chr.5:137605105-137611899 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcolce | NM_008788.2 | NP_032814.2 | ||
| AB008548.1 | BAA23280.1 | |||
| AK142615.1 | BAE25130.1 | |||
| AK144392.1 | BAE25865.1 | |||
| AK146829.1 | BAE27464.1 | |||
| AK159430.1 | BAE35077.1 | |||
| AK159940.1 | ||||
| AK162524.1 | ||||
| X57337.1 | CAA40612.1 |
More Information
Set Membership: |
|

Genomic Map