Assay Details
Target Gene Details
Entrez Gene ID: | 100503670 |
Gene Name: | ribosomal protein L5 |
Gene Aliases: |
U21RNA |
Location: |
Chr.5:107900528-107909005 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpl5 | NM_016980.2 | NP_058676.1 | ||
| AK008489.1 | BAB25695.1 | |||
| AK013107.1 | BAB28652.1 | |||
| AK032778.1 | ||||
| AK088036.1 | ||||
| AK152381.1 | BAE31169.1 | |||
| AK166865.1 | BAE39079.1 | |||
| AK169313.1 | BAE41068.1 | |||
| BC026934.1 | AAH26934.1 | |||
| BC083318.1 | AAH83318.1 | |||
| BC091752.1 | AAH91752.1 | |||
| BC106128.1 | AAI06129.1 | |||
| CJ073701.1 |
More Information
Set Membership: |
|

Genomic Map