Assay Details
Target Gene Details
Entrez Gene ID: | 17969 |
Gene Name: | neutrophil cytosolic factor 1 |
Gene Aliases: |
NCF-47K, NOXO2, Ncf-1, p47-phox, p47<phox>, p47phox |
Location: |
Chr.5:134220053-134229625 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncf1 | NM_001286037.1 | 7 | 684 | NP_001272966.1 |
| NM_010876.4 | 7 | 684 | NP_035006.3 | |
| NR_104383.1 | 7 | 684 | ||
| AB002663.1 | 7 | 663 | BAA25649.1 | |
| AK149619.1 | 7 | 685 | ||
| AK149668.1 | 7 | 651 | BAE29014.1 | |
| AK149732.1 | 7 | 685 | BAE29053.1 | |
| AK150945.1 | 7 | 650 | BAE29979.1 | |
| AK151316.1 | 7 | 651 | BAE30297.1 | |
| AK151573.1 | 6 | 510 | BAE30514.1 | |
| AK151593.1 | 7 | 649 | BAE30533.1 | |
| AK152386.1 | 7 | 647 | BAE31174.1 | |
| AK152992.1 | 7 | 651 | ||
| AK162308.1 | 7 | 651 | BAE36845.1 | |
| AK162623.1 | 7 | 655 | ||
| AK170462.1 | 7 | 651 | BAE41812.1 | |
| AK171559.1 | 7 | 650 | BAE42526.1 | |
| BC055836.1 | 7 | 638 | AAH55836.1 | |
| L11455.1 | 7 | 621 | AAA50469.1 |

Genomic Map