Assay Details
Target Gene Details
Entrez Gene ID: | 231887 |
Gene Name: | PDGFA associated protein 1 |
Gene Aliases: |
HASPP28, PAP, PAP1 |
Location: |
Chr.5:145128770-145140089 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdap1 | NM_001033313.3 | NP_001028485.1 | ||
| AK147170.1 | BAE27734.1 | |||
| BC139204.1 | ||||
| BC139205.1 |

Genomic Map