Assay Details
Target Gene Details
Entrez Gene ID: | 66537 |
Gene Name: | proteasome maturation protein |
Gene Aliases: |
2510048O06Rik, Ump1 |
Location: |
Chr.5:147860628-147875778 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pomp | NM_025624.2 | NP_079900.1 | ||
| AK003439.1 | BAB22791.1 | |||
| AK011115.1 | BAB27410.1 | |||
| AK012455.1 | BAB28250.1 | |||
| AK013934.1 | BAB29067.1 | |||
| AK014296.1 | BAB29252.1 | |||
| AK044283.1 | ||||
| AK167958.1 | BAE39956.1 | |||
| BC027016.1 | AAH27016.1 |
More Information
Set Membership: |
|

Genomic Map