Assay Details
Target Gene Details
Entrez Gene ID: | 117109 |
Gene Name: | processing of precursor 5, ribonuclease P/MRP family (S. cerevisiae) |
Gene Aliases: |
1500019J17Rik, 2700077E03Rik, AW049989, Rnasep3 |
Location: |
Chr.5:115235851-115240972 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pop5 | NM_026398.4 | NP_080674.1 | ||
| AK005290.1 | BAB23935.1 | |||
| AK012421.1 | ||||
| AK013257.1 | BAB28751.1 | |||
| BC022670.1 | AAH22670.1 |
More Information
Set Membership: |
|

Genomic Map