Assay Details
Target Gene Details
Entrez Gene ID: | 74097 |
Gene Name: | processing of precursor 7, ribonuclease P family, (S. cerevisiae) |
Gene Aliases: |
0610037N12Rik, AI852017, Rpp20 |
Location: |
Chr.5:137501439-137502429 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pop7 | NM_028753.2 | NP_083029.1 | ||
| AK002782.1 | BAB22355.1 | |||
| AK162268.1 | BAE36826.1 | |||
| BC010780.1 | AAH10780.1 |
More Information
Set Membership: |
|

Genomic Map