Assay Details
Target Gene Details
Entrez Gene ID: | 18590 |
Gene Name: | platelet derived growth factor, alpha |
Gene Aliases: |
- |
Location: |
Chr.5:138976381-138999916 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdgfa | NM_008808.3 | NP_032834.1 | ||
| AK017169.1 | BAB30622.1 | |||
| AK142653.1 | BAE25146.1 | |||
| AY324648.1 | AAP88590.1 | |||
| BC003817.1 | AAH03817.1 | |||
| M29464.1 | AAA39903.1 |
More Information
Set Membership: |
|

Genomic Map