Assay Details
Target Gene Details
Entrez Gene ID: | 231805 |
Gene Name: | paired immunoglobin-like type 2 receptor alpha |
Gene Aliases: |
AV021745, FDF03 |
Location: |
Chr.5:137821952-137836278 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pilra | NM_153510.3 | NP_705730.1 | ||
| AK041413.1 | BAC30935.1 | |||
| AK041670.1 | ||||
| AK170000.1 | BAE41505.1 |
More Information
Set Membership: |
|

Genomic Map