Assay Details
Target Gene Details
Entrez Gene ID: | 19248 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 12 |
Gene Aliases: |
P19-PTP, PTP-P19, PTP-PEST, PTPG1 |
Location: |
Chr.5:20986645-21055797 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptpn12 | NM_011203.2 | NP_035333.2 | ||
| AK154107.1 | BAE32381.1 | |||
| AK160218.1 | 1 | 122 | BAE35696.1 | |
| BC051980.1 | AAH51980.1 | |||
| X63440.1 | CAA45037.1 |

Genomic Map