Assay Details
Target Gene Details
Entrez Gene ID: | 17120 |
Gene Name: | MAD1 mitotic arrest deficient 1-like 1 |
Gene Aliases: |
AI173502, AW550425, Mad1 |
Location: |
Chr.5:140008689-140321591 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mad1l1 | NM_010752.3 | NP_034882.2 | ||
| AF083812.1 | AAD24499.1 | |||
| AF083813.1 | AAD24500.1 | |||
| AK136981.1 | ||||
| AK169976.1 | BAE41492.1 | |||
| BC010702.1 | AAH10702.1 |

Genomic Map