Assay Details
Target Gene Details
Entrez Gene ID: | 23961 |
Gene Name: | 2'-5' oligoadenylate synthetase 1B |
Gene Aliases: |
Flv, L1, Mmu-L1, Oas1, Oias-2, Oias2, Wnv |
Location: |
Chr.5:120812635-120824163 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oas1b | NM_001083925.1 | 3 | 575 | NP_001077394.1 |
| NR_003507.1 | 3 | 578 | ||
| AB067529.1 | 3 | 578 | BAB84129.1 | |
| AF328926.1 | 3 | 575 | AAM47542.1 | |
| AF418004.1 | 3 | 575 | AAM47544.1 | |
| AF418005.1 | 3 | 575 | AAM47545.1 | |
| AF418006.1 | 3 | 575 | AAM47546.1 | |
| AF418007.1 | 3 | 575 | AAM47547.1 | |
| AF418008.1 | 3 | 575 | AAM47548.1 | |
| AF418009.1 | 3 | 575 | AAM47549.1 | |
| AF466822.1 | 3 | 532 | AAM97603.1 | |
| AF466823.1 | 3 | 532 | AAM97604.1 | |
| AK080152.1 | 3 | 568 | ||
| BC012877.1 | 3 | 567 | AAH12877.1 | |
| X55982.1 | 2 | 341 | CAA39455.1 |
More Information
Set Membership: |
|

Genomic Map