Assay Details
Target Gene Details
Entrez Gene ID: | 26433 |
Gene Name: | procollagen-lysine, 2-oxoglutarate 5-dioxygenase 3 |
Gene Aliases: |
AI414586, LH3 |
Location: |
Chr.5:136987019-136996646 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plod3 | NM_011962.3 | NP_036092.1 | ||
| AF046783.1 | AAD54618.1 | |||
| AK013195.1 | BAB28704.1 | |||
| AK033360.1 | BAC28246.1 | |||
| AK053320.1 | ||||
| AK088948.1 | BAC40669.1 | |||
| AK157741.1 | ||||
| BC043047.1 | AAH43047.1 | |||
| BC054734.1 | AAH54734.1 |
More Information
Set Membership: |
|

Genomic Map