Assay Details
Target Gene Details
Entrez Gene ID: | 215160 |
Gene Name: | rhomboid domain containing 2 |
Gene Aliases: |
0610011L13Rik, AI480532, Rhbdl7, Usmg1 |
Location: |
Chr.5:135632654-135646376 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rhbdd2 | NM_146002.2 | 3 | 788 | NP_666114.1 |
| AK030434.1 | 4 | 844 | ||
| AK032400.1 | 2 | 212 | ||
| AK041845.1 | 3 | 734 | BAC31081.1 | |
| AK043674.1 | 3 | 723 | ||
| AK140006.1 | 3 | 826 | ||
| AK141182.1 | 4 | 909 | ||
| AK161545.1 | 1 | 2674 | BAE36454.1 | |
| AK165926.1 | 3 | 752 | BAE38463.1 | |
| BC018360.1 | 3 | 732 | AAH18360.1 | |
| BC098493.1 | 2 | 442 | AAH98493.1 |
More Information
Set Membership: |
|

Genomic Map