Assay Details
Target Gene Details
Entrez Gene ID: | 56873 |
Gene Name: | limb region 1 |
Gene Aliases: |
1110048D14Rik, AU017641 |
Location: |
Chr.5:29229802-29378397 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lmbr1 | NM_020295.3 | NP_064691.2 | ||
| AF190665.1 | AAF91092.1 | |||
| AK030631.1 | ||||
| AK195974.1 | ||||
| BC016110.1 | AAH16110.1 |
More Information
Set Membership: |
|

Genomic Map