Assay Details
Target Gene Details
Entrez Gene ID: | 17906 |
Gene Name: | myosin, light polypeptide 2, regulatory, cardiac, slow |
Gene Aliases: |
MLC-2, MLC-2v, Mlc2v, Mylpc |
Location: |
Chr.5:122100904-122113396 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myl2 | NM_010861.3 | NP_034991.3 | ||
| AK002367.1 | BAB22045.1 | |||
| AK146674.1 | BAE27350.1 | |||
| M91602.1 | AAA39796.1 |
More Information
Set Membership: |
|

Genomic Map