Assay Details
Target Gene Details
Entrez Gene ID: | 231327 |
Gene Name: | phosphoribosyl pyrophosphate amidotransferase |
Gene Aliases: |
5730454C12Rik, AA408689, AA675351, AI507113, C79945 |
Location: |
Chr.5:76913249-76951578 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppat | NM_172146.2 | 7 | 1308 | NP_742158.1 |
| AK147750.1 | 7 | 958 | BAE28114.1 | |
| AK148034.1 | 7 | 912 | BAE28301.1 | |
| AK159141.1 | 8 | 1458 | ||
| AK159859.1 | 8 | 1458 | ||
| AK160542.1 | 7 | 933 | BAE35859.1 | |
| AK164506.1 | 7 | 922 | BAE37815.1 | |
| AK167057.1 | 7 | 947 | BAE39220.1 | |
| BC023841.1 | 7 | 935 | AAH23841.1 |
More Information
Set Membership: |
|

Genomic Map