Assay Details
Target Gene Details
Entrez Gene ID: | 80291 |
Gene Name: | Rab interacting lysosomal protein-like 2 |
Gene Aliases: |
- |
Location: |
Chr.5:124463265-124478235 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rilpl2 | NM_030259.1 | 2 | 532 | NP_084535.1 |
| AK133760.1 | 2 | 664 | BAE21826.1 | |
| AK150123.1 | 2 | 636 | BAE29324.1 | |
| AK150252.1 | 2 | 635 | BAE29414.1 | |
| BC003324.1 | 2 | 532 | AAH03324.1 |

Genomic Map