Assay Details
Target Gene Details
Entrez Gene ID: | 242819 |
Gene Name: | RUN domain containing 3B |
Gene Aliases: |
Gm440 |
Location: |
Chr.5:8490336-8623027 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rundc3b | NM_198620.1 | NP_941022.1 | ||
| BC057375.1 | AAH57375.1 | |||
| BC058808.1 | AAH58808.1 |

Genomic Map