Assay Details
Target Gene Details
Entrez Gene ID: | 100316903 |
Gene Name: | predicted gene 20605 |
Gene Aliases: |
Lrch4-sap25 |
Location: |
Chr.5:137629123-137642896 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm20605 | NR_033148.1 | |||
| AK164339.1 |
Target Gene Details
Entrez Gene ID: | 751865 |
Gene Name: | sin3 associated polypeptide |
Gene Aliases: |
- |
Location: |
Chr.5:137641473-137642902 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sap25 | NM_001081962.2 | NP_001075431.2 | ||
| AM275337.1 |
More Information
Set Membership: |
|

Genomic Map